Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Dimethoxymethyl)-1,6-naphthyridine 95+%

Supplier: APOLLO SCIENTIFIC
Warning

OR4773-1G OR4773-250MG
77409-988EA 130.72 USD InStock
77409-988 77409-990
2-(Dimethoxymethyl)-1,6-naphthyridine 95+%
2-(Dimethoxymethyl)-1,6-naphthyridine


Formula: C₁₁H₁₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00202920
CAS Number: 386715-36-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty