Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl-2-(2-nitrophenyl)malonate 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR15334-5G OR15334-1G
77405-374EA 243.5 USD InStock
77405-374 77405-372
Dimethyl-2-(2-nitrophenyl)malonate 95%
Dimethyl-2-(2-nitrophenyl)malonate


Formula: C₁₁H₁₁NO₆
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD03617780
CAS Number: 26465-37-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty