Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Nitro-2H-1,4-benzoxazin-3(4H)-one 98%

Supplier: APOLLO SCIENTIFIC

OR110726-1G OR110726-25G OR110726-5G
77403-456EA 29.54 USD InStock
77403-456 77403-458 77403-460
6-Nitro-2H-1,4-benzoxazin-3(4H)-one 98%
6-Nitro-2H-1,4-benzoxazin-3(4H)-one


Formula: C₈H₆N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD03425744
CAS Number: 81721-87-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty