Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-(trifluoromethyl)pyrimidine-5-carbonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC2076J-1G PC2076J-250MG PC2076J-5G
77413-514EA 37.7 USD InStock
77413-514 77413-516 77413-518
2-Chloro-4-(trifluoromethyl)pyrimidine-5-carbonyl chloride 97%
2-Chloro-4-(trifluoromethyl)pyrimidine-5-carbonyl chloride


Formula: C₆HCl₂F₃N₂O
MW: 244.99 g/mol
Boiling Pt: 54 °C/0.15 mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00173789
CAS Number: 154934-99-3
UN: 1760
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty