Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1,1-Trifluoro-2,3-epoxypropane 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC7296-100G PC7296-25G PC7296-5G
77417-778EA 756.98 USD InStock
77417-778 77417-780 77417-782
1,1,1-Trifluoro-2,3-epoxypropane 98%
1,1,1-Trifluoro-2,3-epoxypropane


Formula: C₃H₃F₃O
Boiling Pt: 39.1…39.3 °C/748 mmHg
Melting Pt: –80 °C
Density: 1.33 g/cm³ (25 °C)
Flash Pt: –25 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041506
CAS Number: 359-41-1
UN: 1992
ADR: 3+6.1,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty