Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4,4-Trifluorobutyraldehyde hydrate

Supplier: APOLLO SCIENTIFIC
Warning

PC53109-10G PC53109-1G PC53109-5G
77416-628EA 578.68 USD InStock
77416-628 77416-630 77416-632
4,4,4-Trifluorobutyraldehyde hydrate
4,4,4-Trifluorobutyraldehyde hydrate


Formula: C₄H₇F₃O₂
MW: 144.09 g/mol
Storage Temperature: Refrigerator
CAS Number: 1448773-75-8
UN: 1993
ADR: 3 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty