Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H,1H-Heptafluoro-n-butyl methacrylate 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC4412-100G PC4412-25G PC4412-5G
77415-404EA 635.74 USD InStock
77415-404 77415-406 77415-408
1H,1H-Heptafluoro-n-butyl methacrylate 97%
1H,1H-Heptafluoro-n-butyl methacrylate


Formula: C₈H₇F₇O₂
Boiling Pt: 134…136 °C
Density: 1.345 g/cm³ (25 °C)
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00039251
CAS Number: 13695-31-3
EINECS: 237-217-7
UN: 3272
ADR: 3,III
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty