Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bromodifluoroacetic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC5574-100G PC5574-25G PC5574-5G
77416-794EA 131.42 USD InStock
77416-794 77416-796 77416-798
Bromodifluoroacetic acid 97%
Bromodifluoroacetic acid


Formula: C₂HBrF₂O₂
Boiling Pt: 134 °C
Melting Pt: 26.5 °C
Storage Temperature: Ambient
MDL Number: MFCD02183519
CAS Number: 354-08-5
UN: 3261
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty