Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoropropionic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC1057-1G PC1057-250MG
77412-930EA 45.84 USD InStock
77412-930 77412-932
2-Fluoropropionic acid 97%
2-Fluoropropionic acid


Formula: C₃H₅FO₂
Boiling Pt: 66…67 °C/30 mmHg
Density: 1.181 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD06247714
CAS Number: 6087-13-4
UN: 3265
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty