Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Pyrimidin-2-yl)benzaldehyde 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR6164-1G OR6164-250MG OR6164-5G
77410-780EA 241.46 USD InStock
77410-780 77410-782 77410-784
3-(Pyrimidin-2-yl)benzaldehyde 95%
3-(Pyrimidin-2-yl)benzaldehyde


Formula: C₁₁H₈N₂O
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD08056283
CAS Number: 263349-22-0
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty