Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Chloromethyl)-1-methyl-1H-pyrazole 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR2055-1G OR2055-250MG
77406-562EA 252.66 USD InStock
77406-562 77406-564
3-(Chloromethyl)-1-methyl-1H-pyrazole 97%
3-(Chloromethyl)-1-methyl-1H-pyrazole


Formula: C₅H₇ClN₂
MW: 130.58 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08690275
CAS Number: 84547-64-8
UN: 3265
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty