Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyano-5-methylbenzenesulfonyl chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR25992-1G OR25992-250MG OR25992-5G
77407-718EA 305.64 USD InStock
77407-718 77407-720 77407-722
2-Cyano-5-methylbenzenesulfonyl chloride 98%
2-Cyano-5-methylbenzenesulfonyl chloride


Formula: C₈H₆ClNO₂S
MW: 215.66 g/mol
Melting Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD03659720
CAS Number: 197960-31-9
UN: 3261
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty