Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrahydro-2H-pyran-4-ylacetyl chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR14881-1G OR14881-250MG
77405-130EA 273.04 USD InStock
77405-130 77405-132
Tetrahydro-2H-pyran-4-ylacetyl chloride 95%
Tetrahydro-2H-pyran-4-ylacetyl chloride


Formula: C₇H₁₁ClO₂
Boiling Pt: 110…111 °C/11 mmHg
Storage Temperature: Ambient
MDL Number: MFCD08669649
CAS Number: 40500-05-8
UN: 3265
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty