Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-3-(1,1-difluoroethyl)benzene 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC520586-5G PC520586-1G PC520586-25G
77416-514EA 275.08 USD InStock
77416-514 77416-510 77416-512
1-Bromo-3-(1,1-difluoroethyl)benzene 97%
1-Bromo-3-(1,1-difluoroethyl)benzene


Formula: C₈H₇BrF₂
MW: 221.05 g/mol
Density: 1.5 g/cm³ (25 °C)
Flash Pt: 87.8 °C
Storage Temperature: Ambient
MDL Number: MFCD11007736
CAS Number: 445303-70-8
UN: 3082
ADR: 9+EHS III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty