Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-3-Amino-1-benzylpiperidine 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR15666-10G OR15666-1G OR15666-5G
77405-500EA 234.32 USD InStock
77405-500 77405-502 77405-504
(R)-3-Amino-1-benzylpiperidine 97%
(R)-3-Amino-1-benzylpiperidine


Formula: C₁₂H₁₈N₂
Storage Temperature: Ambient
MDL Number: MFCD03093374
CAS Number: 168466-84-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty