Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-(trifluoromethyl)benzoyl chloride 96%

Supplier: APOLLO SCIENTIFIC
Danger

PC49644-1G PC49644-25G PC49644-5G
77415-942EA 21.34 USD InStock
77415-942 77415-944 77415-946
2-Chloro-6-(trifluoromethyl)benzoyl chloride 96%
2-Chloro-6-(trifluoromethyl)benzoyl chloride


Formula: C₈H₃Cl₂F₃O
MW: 243.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09025361
CAS Number: 916420-44-5
UN: 3265
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty