Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-2-methylphenylacetonitrile 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC0888-1G PC0888-25G PC0888-5G
77412-720EA 37.2 USD InStock
77412-720 77412-722 77412-724
3-Fluoro-2-methylphenylacetonitrile 98%
3-Fluoro-2-methylphenylacetonitrile


Formula: C₉H₈FN
MW: 149.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788544
CAS Number: 500912-15-2
UN: 3439
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty