Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-2-fluoro-3-methoxybenzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

PC302650-1G PC302650-5G
77414-230EA 79.46 USD InStock
77414-230 77414-232
6-Chloro-2-fluoro-3-methoxybenzyl bromide
6-Chloro-2-fluoro-3-methoxybenzyl bromide


Formula: C₈H₇BrClFO
Melting Pt: 47…50 °C
Storage Temperature: Ambient
MDL Number: MFCD04115950
CAS Number: 886499-73-6
UN: 3261
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty