Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl methyl sulfone

Supplier: APOLLO SCIENTIFIC

OR1877-10G OR1877-50G
77406-394EA 69.28 USD InStock
77406-394 77406-396
Benzyl methyl sulfone
Benzyl methyl sulfone


Formula: C₈H₁₀O₂S
Melting Pt: 124…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00025074
CAS Number: 3112-90-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty