Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glycidyl-2,2,3,3-tetrafluoropropyl ether 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC6867H-25G PC6867H-5G
77417-580EA 260.82 USD InStock
77417-580 77417-582
Glycidyl-2,2,3,3-tetrafluoropropyl ether 97%
Glycidyl-2,2,3,3-tetrafluoropropyl ether


Formula: C₆H₈O₂F₄
MW: 188.12 g/mol
Boiling Pt: 50…52 °C
Density: 1.327 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00054682
CAS Number: 19932-26-4
UN: 2810
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty