Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid free radical

Supplier: APOLLO SCIENTIFIC
Warning

OR24671-1G OR24671-250MG
77407-526EA 529.78 USD InStock
77407-526 77407-528
2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid free radical
2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid


Formula: C₉H₁₄NO₃
Melting Pt: 202 °C
Storage Temperature: Ambient
MDL Number: MFCD00041847
CAS Number: 2154-67-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty