Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Diacetylphloroglucinol 96%

Supplier: APOLLO SCIENTIFIC
Warning

OR22030-1G OR22030-500MG
77406-784EA 674.46 USD InStock
77406-784 77406-786
2,4-Diacetylphloroglucinol 96%
2,4-Diacetylphloroglucinol


Formula: C₁₀H₁₀O₅
Melting Pt: 164…168 °C
Storage Temperature: Freezer
MDL Number: MFCD00100483
CAS Number: 2161-86-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty