Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,2,2-Trifluoroethyl)oxirane

Supplier: APOLLO SCIENTIFIC
Danger

PC10086-5G PC10086-1G PC10086-250MG
77412-774EA 316.86 USD InStock
77412-774 77412-770 77412-772
(2,2,2-Trifluoroethyl)oxirane
(2,2,2-Trifluoroethyl)oxirane


Formula: C₄H₅F₃O
Boiling Pt: 82 °C
Density: 1.2487 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00156048
CAS Number: 407-12-5
UN: 1993
ADR: 3,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty