Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Diazabicyclo[2.2.2]octanebis(sulfur dioxide)adduct 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR310371-1G OR310371-25G OR310371-5G
77409-018EA 21.26 USD InStock
77409-018 77409-020 77409-022
1,4-Diazabicyclo[2.2.2]octanebis(sulfur dioxide)adduct 97%
1,4-Diazabicyclo[2.2.2]octanebis(sulfur dioxide)adduct


Formula: C₆H₁₂N₂O₄S₂
MW: 240.29 g/mol
Melting Pt: 168…172 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22377802
CAS Number: 119752-83-9
UN: 1325
ADR: 4.1 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty