Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-Fluoro-2-methylpropionate 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC2925-1G PC2925-5G
77413-976EA 21.26 USD InStock
77413-976 77413-978 77413-980
Methyl 2-Fluoro-2-methylpropionate 95%
Methyl 2-Fluoro-2-methylpropionate


Formula: C₅H₉FO₂
Boiling Pt: 108…109 °C
Density: 1 g/cm³ (25 °C)
Flash Pt: 22 °C
Storage Temperature: Ambient
MDL Number: MFCD03265444
CAS Number: 338-76-1
UN: 3272
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty