Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fluoroiodomethane 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC56130-25G PC56130-5G PC56130-1G
77416-814EA 2109.98 USD InStock
77416-814 77416-816 77416-812
Fluoroiodomethane 95%
Fluoroiodomethane


Formula: CH₂FI
MW: 159.93 g/mol
Boiling Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD12404952
CAS Number: 373-53-5
UN: 2810
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty