Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-1,6-dimethyl-1H-imidazo[4,5-b]pyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR0518T-100MG OR0518T-10MG OR0518T-5MG
77402-108EA 1994.84 USD InStock
77402-108 77402-110 77402-112
2-Amino-1,6-dimethyl-1H-imidazo[4,5-b]pyridine
2-Amino-1,6-dimethyl-1H-imidazo[4,5-b]pyridine
A mutagenic cogener of PHIP.


Formula: C₈H₁₀N₄
Storage Temperature: Freezer
MDL Number: MFCD00269879
CAS Number: 132898-04-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty