Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitropyridine 95%

Supplier: APOLLO SCIENTIFIC

OR10961-1G OR10961-250MG OR10961-5G
77403-000EA 92.72 USD InStock
77403-000 77403-002 77403-004
4-Nitropyridine 95%
4-Nitropyridine


Formula: C₅H₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00054554
CAS Number: 1122-61-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty