Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-octylbenzoate

Supplier: APOLLO SCIENTIFIC

OR110268-1G OR110268-5G
77403-200EA 121.24 USD InStock
77403-200 77403-202
Ethyl 4-octylbenzoate
Ethyl 4-octylbenzoate


Formula: C₁₇H₂₆O₂
MW: 262.387 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06204568
CAS Number: 133002-71-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty