Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-6-(1H-pyrazol-1-yl)pyrimidine 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR11468-100MG OR11468-1G
77403-926EA 28.52 USD InStock
77403-926 77403-928
4-Chloro-6-(1H-pyrazol-1-yl)pyrimidine 98%
4-Chloro-6-(1H-pyrazol-1-yl)pyrimidine


Formula: C₇H₅ClN₄
Storage Temperature: Refrigerator
MDL Number: MFCD00297274
CAS Number: 114833-95-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty