Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-Chlorophenyl)benzylamine hydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

OR12640-50MG OR12640-1G OR12640-250MG
77404-394EA 69.28 USD InStock
77404-394 77404-390 77404-392
4-(2-Chlorophenyl)benzylamine hydrochloride
4-(2-Chlorophenyl)benzylamine hydrochloride


Formula: C₁₃H₁₂ClN·ClH
Storage Temperature: Ambient
MDL Number: MFCD02089431
CAS Number: 518357-42-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty