Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclohexylmethanesulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR12997-100MG OR12997-1G OR12997-250MG
77404-480EA 210.9 USD InStock
77404-480 77404-482 77404-484
Cyclohexylmethanesulfonyl chloride
Cyclohexylmethanesulfonyl chloride


Formula: C₇H₁₃ClO₂S
MW: 196.695 g/mol
Boiling Pt: 92…95 °C (0.5 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD03452748
CAS Number: 4352-30-1
UN: 3265
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty