Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Aminoethyl)-4-bromo-1-methyl-1H-pyrazole

Supplier: APOLLO SCIENTIFIC
Warning

OR17548-1G OR17548-250MG
77406-036EA 690.76 USD InStock
77406-036 77406-038
5-(Aminoethyl)-4-bromo-1-methyl-1H-pyrazole
5-(Aminoethyl)-4-bromo-1-methyl-1H-pyrazole


Formula: C₆H₁₀BrN₃
Storage Temperature: Refrigerator
MDL Number: MFCD08449281
CAS Number: 1017788-72-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty