Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-2-benzylpropanoic acid, N-BOC protected 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR2225-1G OR2225-250MG OR2225-5G
77406-856EA 210.9 USD InStock
77406-856 77406-858 77406-860
3-Amino-2-benzylpropanoic acid, N-BOC protected 98%
3-Amino-2-benzylpropanoic acid, N-BOC protected


Formula: C₁₅H₂₁NO₄
Melting Pt: 94…102 °C
Storage Temperature: Ambient
MDL Number: MFCD03003690
CAS Number: 26250-90-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty