Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Iodo-7-methoxyisoquinoline

Supplier: APOLLO SCIENTIFIC
Warning

OR300132-100MG OR300132-250MG OR300132-500MG
77408-568EA 66.22 USD InStock
77408-568 77408-570 77408-572
1-Iodo-7-methoxyisoquinoline
1-Iodo-7-methoxyisoquinoline


Formula: C₁₀H₈INO
Melting Pt: 86…87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04037478
CAS Number: 952569-54-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty