Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Bromomethyl)-7-chlorobenzo[b]thiophene 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR31658-1G OR31658-5G
77409-102EA 110.04 USD InStock
77409-102 77409-104
3-(Bromomethyl)-7-chlorobenzo[b]thiophene 98%
3-(Bromomethyl)-7-chlorobenzo[b]thiophene


Formula: C₉H₆BrClS
MW: 261.566 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07787461
CAS Number: 17512-61-7
UN: 3261
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty