Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Dimethyl-4-oxobutanenitrile 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR40004-100MG OR40004-250MG
77409-422EA 256.2 USD InStock
77409-422 77409-424 77409-426
2,2-Dimethyl-4-oxobutanenitrile 95%
2,2-Dimethyl-4-oxobutanenitrile


Formula: C₆H₉NO
Storage Temperature: Refrigerator
MDL Number: MFCD07437928
CAS Number: 18240-73-8
UN: 3276
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty