Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyclopropylbenzenesulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR45173-1G OR45173-250MG OR45173-5G
77409-818EA 121.24 USD InStock
77409-818 77409-820 77409-822
3-Cyclopropylbenzenesulfonyl chloride
3-Cyclopropylbenzenesulfonyl chloride


Formula: C₉H₉ClO₂S
MW: 216.685 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19201052
CAS Number: 958651-12-2
UN: 3261
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty