Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Cyclopropyl)thiophene-2-carbonitrile

Supplier: APOLLO SCIENTIFIC

OR45202-250MG OR45202-1G
77409-834EA 220.08 USD InStock
77409-834 77409-832
4-(Cyclopropyl)thiophene-2-carbonitrile
4-(Cyclopropyl)thiophene-2-carbonitrile


Formula: C₈H₇NS
Storage Temperature: Refrigerator
MDL Number: MFCD28059592
CAS Number: 1823927-97-4
UN: 1993
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty