Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Chlorosulfonyl)azetidine, N-BOC protected

Supplier: APOLLO SCIENTIFIC
Danger

OR46226-1G OR46226-250MG
77409-902EA 536.92 USD InStock
77409-902 77409-904
3-(Chlorosulfonyl)azetidine, N-BOC protected
3-(Chlorosulfonyl)azetidine, N-BOC protected


Formula: C₈H₁₄ClNO₄S
Storage Temperature: Freezer
MDL Number: MFCD11040529
CAS Number: 1310732-18-3
UN: 1759
ADR: 8,III
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty