Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Bromomethyl)benzyl alcohol 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR52377-10G OR52377-1G OR52377-5G
77410-236EA 348.44 USD InStock
77410-236 77410-238 77410-240
4-(Bromomethyl)benzyl alcohol 97%
4-(Bromomethyl)benzyl alcohol


Formula: C₈H₉BrO
MW: 201.06 g/mol
Melting Pt: 76…77 °C
Storage Temperature: Freezer
MDL Number: MFCD08275218
CAS Number: 71831-21-5
UN: 3261
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty