Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dimethoxy-2-hydroxyphenacyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

OR54698-1G OR54698-250MG OR54698-5G
77410-428EA 23.34 USD InStock
77410-428 77410-430 77410-432
3,4-Dimethoxy-2-hydroxyphenacyl bromide
3,4-Dimethoxy-2-hydroxyphenacyl bromide


Formula: C₁₀H₁₁BrO₄
MW: 275.096 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32177107
CAS Number: 18064-92-1
UN: 3262
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty