Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethyl)cinnamamide 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC0552-1G PC0552-5G
77412-626EA 22.36 USD InStock
77412-626 77412-628
4-(Trifluoromethyl)cinnamamide 98%
4-(Trifluoromethyl)cinnamamide


Formula: C₁₀H₈F₃NO
Storage Temperature: Refrigerator
MDL Number: MFCD03092988
CAS Number: 115093-99-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty