Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dichloro-2-fluoroaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC14291-100G PC14291-25G PC14291-50G
77413-180EA 175.24 USD InStock
77413-180 77413-182 77413-184
3,4-Dichloro-2-fluoroaniline 98%
3,4-Dichloro-2-fluoroaniline


Formula: C₆H₄Cl₂FN
Melting Pt: 53…56 °C
Density: 1.5 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD04039241
CAS Number: 886762-39-6
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty