Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Dimethyl-4,4,4-trifluorobutanoic acid 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC1824-1G PC1824-250MG
77413-332EA 574.64 USD InStock
77413-332 77413-334
2,2-Dimethyl-4,4,4-trifluorobutanoic acid 95%
2,2-Dimethyl-4,4,4-trifluorobutanoic acid


Formula: C₆H₉F₃O₂
Boiling Pt: 86…87 °C (6 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09998070
CAS Number: 939399-07-2
UN: 3265
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty