Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Difluoromethoxy)benzenesulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

PC2382-5G PC2382-25G
77413-614EA 228.02 USD InStock
77413-614 77413-612
3-(Difluoromethoxy)benzenesulfonyl chloride
3-(Difluoromethoxy)benzenesulfonyl chloride


Formula: C₇H₅ClF₂O₃S
MW: 242.628 g/mol
Boiling Pt: 233…234 °C
Density: 1.509 g/cm³ (25 °C)
Flash Pt: 97.8 °C
Storage Temperature: Ambient
MDL Number: MFCD03094157
CAS Number: 351003-38-8
UN: 3265
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty