Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2-fluoro-2-methylpropane 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC2438-1G PC2438-500MG
77413-650EA 227.2 USD InStock
77413-650 77413-652
1-Bromo-2-fluoro-2-methylpropane 95%
1-Bromo-2-fluoro-2-methylpropane


Formula: C₄H₈BrF
MW: 155.009 g/mol
Boiling Pt: 95…96 °C
Storage Temperature: Ambient
MDL Number: MFCD04972692
CAS Number: 19869-78-4
UN: 1993
ADR: 3 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty