Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethoxy-1,1,1-trifluoro-3-methylbut-3-en-2-one

Supplier: APOLLO SCIENTIFIC
Warning

PC28305-1G PC28305-250MG PC28305-5G
77413-898EA 573.48 USD InStock
77413-898 77413-900 77413-902
4-Ethoxy-1,1,1-trifluoro-3-methylbut-3-en-2-one
4-Ethoxy-1,1,1-trifluoro-3-methylbut-3-en-2-one


Formula: C₇H₉F₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD30730450
CAS Number: 153085-12-2
UN: 1993
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty