Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-5-fluorobenzoyl chloride

Supplier: APOLLO SCIENTIFIC

PC302400-10G PC302400-5G
77414-164EA 344.36 USD InStock
77414-164 77414-168
3-Chloro-5-fluorobenzoyl chloride
3-Chloro-5-fluorobenzoyl chloride


Formula: C₇H₃Cl₂FO
MW: 193.003 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115835
CAS Number: 886496-62-4
UN: 3265
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty