Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-5-(trifluoromethoxy)benzenesulfonyl chloride 97%

Supplier: APOLLO SCIENTIFIC

PC303230-1G PC303230-250MG
77414-374EA 853.76 USD InStock
77414-374 77414-376
2-Methyl-5-(trifluoromethoxy)benzenesulfonyl chloride 97%
2-Methyl-5-(trifluoromethoxy)benzenesulfonyl chloride


Formula: C₈H₆ClF₃O₃S
MW: 274.645 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18399733
CAS Number: 1261852-46-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty